Learn More
c-Myc Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody has been used in 45 publications
Brand:  Novus Biologicals NB600-335
Product Code. 18788473
  • ONLINE EXCLUSIVE PRICE
    € 509.00
    € 480.90 / 0.10mL
    Save € 28.10
    5% Off
Description
c-Myc Polyclonal specifically detects c-Myc in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
0.1 mL
IgG
Goat
Store at 4C. Do not freeze.