missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-336IR
This item is not returnable.
View return policy
Description
c-Myc Polyclonal antibody specifically detects c-Myc in Human, Mouse, Insect, S. pombe samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
| c-Myc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Insect, S. pombe | |
| Purified |
| Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DyLight 755 | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| A synthetic peptide made to the human c-Myc Antibody (between residues 385-435) [Uniprot: P01106] | |
| 0.1 mL | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 4609 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction