missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (MYC/7150R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20786AF405
This item is not returnable.
View return policy
Description
c-Myc Monoclonal antibody specifically detects c-Myc in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| c-Myc | |
| Monoclonal | |
| Alexa Fluor 405 | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Recombinant fragment (around aa1-200) of human c-Myc protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 4609 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MYC/7150R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction