missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (Myc.A7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37822AF350
This item is not returnable.
View return policy
Description
c-Myc Monoclonal specifically detects c-Myc in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| c-Myc | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MYC | |
| Synthetic peptide: EQKLISEEDL c-Myc Antibody (Myc.A7). | |
| 0.1 mL | |
| Primary | |
| Detects N-terminal or C-terminal Myc tagged fusion proteins. It has been successfully used in ELISA, immunofluorescence, immunoprecipitation and Western Blot procedures. immunogen is a synthetic peptide EQKLISEEDL (Myc). | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Myc.A7 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Mouse | |
| 63 kDa | |
| Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 4609 | |
| Non-species specific | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction