missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Antibody (CC27) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-05434JF635
This item is not returnable.
View return policy
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Rat samples. It is validated for ELISA,Immunoassay,Sandwich ELISA
Specifications
| C-Peptide | |
| Monoclonal | |
| Janelia Fluor 635 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of rat C-peptides I and II conjugated with the carrier protein. | |
| 0.2 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunoassay, Sandwich ELISA | |
| CC27 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction