missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Monoclonal Antibody (C-PEP-01), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110317
This item is not returnable.
View return policy
Description
After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into three peptides: the B chain and A chain peptides, which are covalently linked via two disulfide bonds to form insulin, and C-peptide. Binding of insulin to the insulin receptor (INSR) stimulates glucose uptake. A multitude of mutant alleles with phenotypic effects have been identified.Specifications
C-Peptide | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01308 | |
INS | |
Human C-peptide conjugated to bovine serum albumin. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
C-PEP-01 | |
Biotin | |
INS | |
Cpeptide; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2-rs1; insulin; insulin 1; Insulin A chain; Insulin B chain; insulin I; insulin-1; Insulin-1 A chain; Insulin-1 B chain; IRDN; MODY10; preproinsulin; proinsulin | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3630 | |
4° C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction