missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Reactive Protein/CRP Antibody (C7), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1333AF532
This item is not returnable.
View return policy
Description
C-Reactive Protein/CRP Monoclonal antibody specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, ELISA, Affinity Purification
Specifications
| C-Reactive Protein/CRP | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunoassay, ELISA, Affinity Purification | |
| C7 | |
| ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification | |
| C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human CRP. | |
| 0.2 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1401 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction