missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C10orf32 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15725931 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15725931 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15725931 Supplier Invitrogen™ Supplier No. PA558040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FGQSVKGLLT EKVTTCGTDV IALTKQVLKG SRSSELLGQA ARNMVLQEDA ILHSEDSLRK MAIITTHLQY QQEAIQKNVE QSSDLQDQLN HLLK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

C10orf32 (chromosome 10 open reading frame 32) is a 105 amino acid protein that belongs to the UPF0693 family. The gene encoding C10orf32 maps to human chromosome 10, which spans nearly 135 million base pairs, makes up approximately 4.5% of total DNA in cells and encodes nearly 1,200 genes. Several protein-coding genes, including those that encode for chemokines, cadherins, excision repair proteins, early growth response factors (Egrs) and fibroblast growth receptors (FGFRs), are located on chromosome 10. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie-Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman's syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C10orf32
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BORCS7
Gene Accession No. Q96B45
Gene Alias 2010012O05Rik; 4930569L17Rik; AI413851; AI467257; BLOC-1 related complex subunit 7; BLOC-1-related complex subunit 7; BORCS7; C10orf32; diaskedin; RGD1311783; RIKEN cDNA 2010012O05 gene; similar to RIKEN cDNA 2010012O05; UPF0693 protein C10orf32; UPF0693 protein C10orf32 homolog
Gene Symbols BORCS7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C10orf32. Recombinant protein control fragment (Product #RP-97173).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.