missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C10orf4 Polyclonal Antibody

Description
Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Human (99%), Mouse (90%), Rat (91%), Pig (93%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a nuclear phosphoprotein of unknown function. The 5'; UTR of this gene is part of a CpG island and contains a tandem CGG repeat region that normally consists of 8-14 repeats but can expand to over 200 repeats. The expanded allele becomes hypermethylated and is not transcribed; however, an expanded repeat region has not been associated with any disease phenotype. This gene is found within the rare FRA10A folate-sensitive fragile site.
Specifications
Specifications
| Antigen | C10orf4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | FRA10AC1 |
| Gene Accession No. | Q70Z53 |
| Gene Alias | 5730455O13Rik; AI448293; C10orf4; F26C11.1-like; FRA10A; Fra10ac1; FRA10AC1 homolog (human); fragile site 10q23.3; fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1; PRO2972; Protein FRA10AC1; protein FRA10AC1 homolog; rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1; RGD1309482 |
| Gene Symbols | FRA10AC1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?