Learn More
Invitrogen™ C10orf46 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113995
Description
Immunogen sequence: SELSEYAAQD QKFQRELIQN GFTRGDQSRK RAGDELAYNS SSACASSRGY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.
Specifications
| C10orf46 | |
| Polyclonal | |
| Unconjugated | |
| CACUL1 | |
| 2700078E11Rik; 2810417M16Rik; 9830127L17Rik; AI450346; C10orf46; CAC1; CACUL1; CDK2 associated cullin domain 1; CDK2 associated, cullin domain 1; CDK2-associated and cullin domain-containing protein 1; CDK2-associated, cullin domain 1; Cdk-associated cullin1; cullin; D130033C15Rik; RGD1308127 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 143384 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86Y37 | |
| CACUL1 | |
| Synthetic peptide directed towards the middle region of human C10orf46. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.