missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf54 Monoclonal Antibody (OTI1D10C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812262S
This item is not returnable.
View return policy
Description
C10orf54 Monoclonal antibody specifically detects C10orf54 in Human samples. It is validated for Western BlotSpecifications
| C10orf54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14784 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3560 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1D10C1 | |
| Unconjugated | |
| Il2rb | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| Human recombinant protein fragment corresponding to amino acids 27-240+Mouse FC of human IL2RB produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction