missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf54 Monoclonal Antibody (OTI7E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812250
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| C10orf54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| B7-H5, B7H5, C10orf54, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64115 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI7E9 | |
| Unconjugated | |
| VSIR | |
| VSIR | |
| Full length human recombit protein of human C10ORF54 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction