missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf82 Antibody (OTI1G4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72401R
This item is not returnable.
View return policy
Description
C10orf82 Monoclonal specifically detects C10orf82 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| C10orf82 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C10orf82 | |
| Full length recombinant protein of human C10orf82 (NP_653262) produced in HEK293T cells. | |
| 0.1 mL | |
| 143379.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1G4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| chromosome 10 open reading frame 82, Em:AC016825.4, FLJ40268, hypothetical protein LOC143379, MGC33547 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction