missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf67 Monoclonal Antibody (OTI4C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803536S
This item is not returnable.
View return policy
Description
C11orf67 Monoclonal antibody specifically detects C11orf67 in Human samples. It is validated for Western BlotSpecifications
| C11orf67 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P12644 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 652 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI4C1 | |
| Unconjugated | |
| BMP4 | |
| 1810020D17Rik; 1810037D19Rik; Aamdc; adipogenesis associated Mth938 domain containing; adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; C11orf67; C29H11orf67; CK067; hypothetical protein LOC553802; LI2; Mth938 domain-containing protein; PTD015; RGD1561459; Unknown (protein for MGC:133476); UPF0366 protein C11orf67; UPF0366 protein C11orf67 homolog; zgc:112239 | |
| Human recombinant protein fragment corresponding to amino acids 293-408 of human BMP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction