missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C11orf91 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15988204
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15988204 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15988204 Supplier Invitrogen™ Supplier No. PA565463

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SGLASIPYEPL RFFYSPPPGP EVVASPLVPC PSTPRLASAS HPEELCELEI Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

C11orf91 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C11orf91
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C11orf91
Gene Accession No. Q3C1V1
Gene Alias C11orf91; chromosome 11 open reading frame 91; uncharacterized protein C11orf91
Gene Symbols C11orf91
Host Species Rabbit
Immunogen Recombinant Human C11orf91. Recombinant protein control fragment (Product #RP-106141).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.