missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C12orf29 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92281-0.02ml
This item is not returnable.
View return policy
Description
C12orf29 Polyclonal antibody specifically detects C12orf29 in Human, Mouse samples. It is validated for Western Blot
Specifications
| C12orf29 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| chromosome 12 open reading frame 29, DKFZp313K0436, DKFZp434N2030, DKFZp686L04169, FLJ38158, hypothetical protein LOC91298, MGC102978 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-325 of human C12orf29 (NP_001009894.2). MKRLGSVQRKMPCVFVTEVKEEPSSKREHQPFKVLATETVSHKALDADIYSAIPTEKVDGTCCYVTTYKDQPYLWARLDRKPNKQAEKRFKNFLHSKENPKEFFWNVEEDFKPAPECWIPAKETEQINGNPVPDENGHIPGWVPVEKNNKQYCWHSSVVNYEFEIALVLKHHPDDSGLLEISAVPLSDLLEQTLELIGTNINGNPYGLGSKKHPLHLLIPHGAFQIRNLPSLKHNDLVSWFEDCKEGKIEGIVWHCSDGCLIKVHRHHLGLCWPIPDTYMNSRPVIINMNLNKCDSAFDIKCLFNHFLKIDNQKFVRLKDIIFDV | |
| 0.02 mL | |
| Cell Biology | |
| 91298 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction