missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C12orf4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99258B
This item is not returnable.
View return policy
Description
C12orf4 Polyclonal antibody specifically detects C12orf4 in Human samples. It is validated for Western Blot
Specifications
| C12orf4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57102 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| chromosome 12 open reading frame 4, hypothetical protein LOC57102 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C12orf4. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction