missing translation for 'onlineSavingsMsg'
Learn More

C12orf49 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15733381 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15733381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15733381 Supplier Thermo Scientific Supplier No. PA555459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C12orf49 Polyclonal Antibody

C12orf49 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf49
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C12orf49
Gene Accession No. Q9H741
Gene Alias C12orf49; chromosome 12 open reading frame 49; UPF0454 protein C12orf49
Gene Symbols C12orf49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C12orf49. Recombinant protein control fragment (Product #RP-90102).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.