missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C12orf60 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18035681
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035681 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18035681 Supplier Invitrogen™ Supplier No. PA5144799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EKDKERLIQA AKMFFFHVQD LASVINTLTE LFSRSMNTQI LLMAVKNNSY IKDFFEQMLR IFKEMQSVVD ARHDKIQKES LCSKV Antigen sequence identity: Mouse 66%; Rat 58%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C12orf60
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C12orf60
Gene Accession No. Q5U649
Gene Alias C12orf60; chromosome 12 open reading frame 60; uncharacterized protein C12orf60
Gene Symbols C12orf60
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 6-90. Recombinant protein control fragment (Product #RP-110041).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144608
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.