missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C13orf15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17731974
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17731974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17731974 Supplier Invitrogen™ Supplier No. PA5120573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, Mouse kidney, Mouse heart. Immunogen sequence: ADFASPFHER HFHYEEHLER MKRRSSASVS DSSGFSDSES ADSLYRNSFS FSDEKLNSPT D.

Modulates the activity of cell cycle-specific kinases. Enhances CDK1 activity. May contribute to the regulation of the cell cycle. May inhibit growth of glioma cells by promoting arrest of mitotic progression at the G2/M transition. Fibrogenic factor contributing to the pathogenesis of renal fibrosis through fibroblast activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C13orf15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene RGCC
Gene Accession No. Q9DBX1, Q9H4X1, Q9Z2P4
Gene Alias 1190002H23Rik; bA157L14.2; C13orf15; regulator of cell cycle; regulator of cell cycle RGCC; Response gene to complement 32 protein; Rgc32; RGC-32; Rgcc
Gene Symbols RGCC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 40-100 of human RGCC (NP_054778.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117183, 28984, 66214
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.