Learn More
C13orf39 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26260-1-AP-20UL
 View more versions of this product
Product Code. 18058775
  • € 145.00 / 20µL
Description
The C13orf39 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C13orf39. This antibody recognizes human, rat antigen. The C13orf39 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
C13orf39
1000 μg/mL
Rabbit
Primary
Human, Rat
150 kDa
IgG
Western Blot, ELISA
C13orf39, UPF0567 protein C13orf39
20 μL
196541
Store at -20°C. Stable for one year after shipment.