Learn More
C13orf39 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26260-1-AP-20UL
Description
The C13orf39 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C13orf39. This antibody recognizes human, rat antigen. The C13orf39 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| C13orf39 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| C13orf39, UPF0567 protein C13orf39 | |
| 20 μL | |
| 196541 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |