missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551469
This item is not returnable.
View return policy
Description
Immunogen sequence: RQSELEPVVS LVDVLEEDEE LENEACAVLG GSDSEKCSYS QGSVKRQALY ACSTCTPEGE EPAGICLACS YECHGSHKLF ELYTKRNFRC DCGNSKFKNL ECKLLPDKAK VNSGNKYNDN FFGLYCICKR PYPDPEDEI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway, recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
| C14orf130 | |
| Polyclonal | |
| Unconjugated | |
| UBR7 | |
| 5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N806 | |
| UBR7 | |
| Recombinant protein corresponding to Human C14orf130. Recombinant protein control fragment (Product #RP-88891). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction