missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C14orf140 Polyclonal Antibody

Product Code. 15760062 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15760062 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760062 Supplier Invitrogen™ Supplier No. PA562267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ENENGELQKI ILPRSRVKGN KSNTMYKPIF SPEFEFEEEF SRDRREDETW GRSQQNSVPF QFSDYRIQRL KRERLVASNN KIRDP Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 53%.

FAM164C, also named as ZC2HC1C or C14orf140, is a 456 amino acid protein, which contains one C2HC-type zinc finger and belongs to the ZC2HC1 family. FAM164C exists as two isoforms. The isoform1 is a 51 kDa protein ad isoform2 is 31 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf140
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC2HC1C
Gene Accession No. Q53FD0
Gene Alias 2810002I04Rik; AV046379; C14orf140; FAM164C; family with sequence similarity 164, member C; protein FAM164C; RGD1307122; ZC2HC1C; Zinc finger C2HC domain-containing protein 1C; zinc finger C2HC-type containing 1C; zinc finger, C2HC-type containing 1C
Gene Symbols ZC2HC1C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C14orf140. Recombinant protein control fragment (Product #RP-101238).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.