missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf140 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584681
This item is not returnable.
View return policy
Description
Immunogen sequence: SRNFGVRNQG NFSVVGTVLA ATQAEKAVAN FDRTEWVQIR RLEAAGESLE EEIRRKQILL RGKLKKTEEE LRRIQTQK Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 62%.
FAM164C, also named as ZC2HC1C or C14orf140, is a 456 amino acid protein, which contains one C2HC-type zinc finger and belongs to the ZC2HC1 family. FAM164C exists as two isoforms. The isoform1 is a 51 kDa protein ad isoform2 is 31 kDa.
Specifications
| C14orf140 | |
| Polyclonal | |
| Unconjugated | |
| ZC2HC1C | |
| 2810002I04Rik; AV046379; C14orf140; FAM164C; family with sequence similarity 164, member C; protein FAM164C; RGD1307122; ZC2HC1C; Zinc finger C2HC domain-containing protein 1C; zinc finger C2HC-type containing 1C; zinc finger, C2HC-type containing 1C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53FD0 | |
| ZC2HC1C | |
| Recombinant protein corresponding to Human ZC2HC1C. Recombinant protein control fragment (Product #RP-108231). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction