Learn More
C14orf166 Monoclonal Antibody (OTI1G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804052
Encompass_Preferred
 View more versions of this product
Product Code. 17533383
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
The function of the C14orf266 gene has not yet been determined.
Specifications
Specifications
C14orf166
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y224
RTRAF
Full length human recombit protein of human C14orf166 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI1G8
Unconjugated
RTRAF
C14orf166, CGI-99, CGI99, CLE, CLE7, LCRP369, RLLM1, hCLE, hCLE1
Mouse
Affinity Chromatography
RUO
51637
-20° C, Avoid Freeze/Thaw Cycles