missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf166 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804059S
This item is not returnable.
View return policy
Description
C14orf166 Monoclonal antibody specifically detects C14orf166 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| C14orf166 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y224 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51637 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C3 | |
| Unconjugated | |
| RTRAF | |
| 2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog | |
| Full length human recombinant protein of human C14orf166 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction