Learn More
C14orf166 Monoclonal Antibody (OTI2D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804056S
Encompass_Preferred
 View more versions of this product
Product Code. 17543393
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
C14orf166 Monoclonal antibody specifically detects C14orf166 in Human samples. It is validated for Western Blot
Specifications
Specifications
C14orf166
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y224
Mouse
Affinity chromatography
RUO
51637
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2D6
Unconjugated
RTRAF
2700060E02Rik; C14orf166; CGI99; CGI-99; chromosome 14 open reading frame 166; CLE; CLE7; CLE7 homolog; hCLE1; LCRP369; LRRGT00192; RGD1304704; RIKEN cDNA 2700060E02 gene; RLL motif containing 1; RLLM1; RNA transcription, translation and transport factor protein; RTRAF; similar to Hypothetical protein CGI-99; UPF0568 protein C14orf166; UPF0568 protein C14orf166 homolog
Full length human recombinant protein of human C14orf166 produced in E.coli.
30 μL