missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C14orf21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531719
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (80%), Rat (83%), Rhesus Monkey (98%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The C14orf21 gene, also known as SLIRP (SRA stem-loop interacting RNA binding protein), is located on chromosome 14 and encodes a mitochondrial protein involved in RNA processing. SLIRP is crucial for the stabilization and regulation of mitochondrial-encoded RNAs, which play a vital role in mitochondrial gene expression and overall mitochondrial function. The SLIRP protein interacts with RNA stem-loop structures, particularly those involved in the steroid receptor RNA activator (SRA), thereby regulating gene expression by modulating the activity of nuclear receptors. Defects in SLIRP can lead to impaired mitochondrial function, resulting in various clinical manifestations, including mitochondrial myopathy and other disorders related to mitochondrial dysfunction.
Specifications
| C14orf21 | |
| Polyclonal | |
| Unconjugated | |
| NOP9 | |
| C14orf21; KIAA2021; NOP9; NOP9 nucleolar protein; NOP9 nucleolar protein homolog; nucleolar protein 9; pumilio domain-containing protein C14orf21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 161424 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q86U38 | |
| NOP9 | |
| Recombinant protein encompassing a sequence within the center region of human C14orf21. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction