Learn More
Invitrogen™ C15orf2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550617
Product Code. 15664545
  • € 520.00 / 100µL
Description
The antibody detects endogenous levels of total C15orf2 / NPAP1 protein.
May be involved in spermatogenesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C15orf2
Polyclonal
Unconjugated
NPAP1
C15orf2; NPAP1; nuclear pore associated protein 1; nuclear pore-associated protein 1; protein C15orf2
Rabbit
Antigen affinity chromatography
RUO
23742
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9NZP6
NPAP1
Synthetic peptide corresponding to a region derived from internal residues of human nuclear pore associated protein 1.