missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C15orf40 Antibody (OTI2B7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72375IR
This item is not returnable.
View return policy
Description
C15orf40 Monoclonal specifically detects C15orf40 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| C15orf40 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C15orf40 | |
| Full length recombinant protein of human C15orf40 (NP_653198) produced in HEK293T cells. | |
| 0.1 mL | |
| 123207.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI2B7 | |
| Western Blot, Flow Cytometry | |
| chromosome 15 open reading frame 40, FLJ33606, hypothetical protein LOC123207, MGC29937 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction