missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C15orf41 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563922
This item is not returnable.
View return policy
Description
Immunogen sequence: TKAQYDEIAQ CLVSVPPTRQ SLRKLKQRFP SQSQATLLSI FSQEYQKHIK RTHAKHHTSE AIESYYQRYL NGVVKNGAAP VLLDLA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Specifications
| C15orf41 | |
| Polyclonal | |
| Unconjugated | |
| C15orf41 | |
| BC052040; C15orf41; cDNA sequence BC052040; chromosome 15 open reading frame 41; HH114; Protein C15orf41; Protein C15orf41 homolog; Protein HH114; uncharacterized protein C15orf41; uncharacterized protein C15orf41 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84529 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2V0 | |
| C15orf41 | |
| Recombinant protein corresponding to Human C15orf41. Recombinant protein control fragment (Product #RP-101975). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction