missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C15orf48 Polyclonal Antibody
C15orf48 Polyclonal Antibody
Brand: Invitrogen PA569665
This item is not returnable.
View return policy
Description
C15orf48 Polyclonal Antibody for Western Blot
Specifications
| C15orf48 | |
| Polyclonal | |
| Unconjugated | |
| C15orf48 | |
| C15orf48; chromosome 15 open reading frame 48; FOAP-11; NMES1; normal mucosa of esophagus specific 1; normal mucosa of esophagus-specific gene 1 protein; Protein FOAP-11 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84419 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9C002 | |
| C15orf48 | |
| synthetic peptide directed towards the middle region of human C15orf48 (aa 22-71). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction