missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf57 Monoclonal Antibody (OTI4B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811338
This item is not returnable.
View return policy
Description
C16orf57 is a phosphodiesterase responsible for the U6 snRNA 3′ end processing. This protein acts as an exoribonuclease (RNase) responsible for trimming the poly(U) tract of the last nucleotides in the pre-U6 snRNA molecule, leading to the formation of mature U6 snRNA 3′ end-terminated with a 2′,3′-cyclic phosphate.Specifications
| C16orf57 | |
| Monoclonal | |
| Unconjugated | |
| USB1 | |
| C16orf57, HVSL1, Mpn1, PN, hUsb1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4B1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQ65 | |
| USB1 | |
| Full length human recombit protein of human C16orf57 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction