missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf57 Monoclonal Antibody (OTI4B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811338S
This item is not returnable.
View return policy
Description
C16orf57 Monoclonal antibody specifically detects C16orf57 in Human samples. It is validated for Western BlotSpecifications
| C16orf57 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P28749 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5933 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4B1 | |
| Unconjugated | |
| RBL1 | |
| AA960436; C16orf57; FLJ13154; HAMAP-Rule:MF_03040}; hUsb1; HVSL motif containing 1; HVSL1; Mpn1; mutated in poikiloderma with neutropenia protein 1; PN; putative U6 snRNA phosphodiesterase; RGD1305215; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA biogenesis phosphodiesterase 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1 | |
| Human recombinant protein fragment corresponding to amino acids 739-1014 of human RBL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction