missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf72 Antibody (OTI2D1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71842AF350
This item is not returnable.
View return policy
Description
C16orf72 Monoclonal specifically detects C16orf72 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| C16orf72 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| C16orf72 | |
| Full length recombinant protein of human C16orf72 (NP_054836) produced in HEK293T cells. | |
| 0.1 mL | |
| 29035.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2D1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction