missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf28 Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501359
This item is not returnable.
View return policy
Description
CQ028 may play an important role in the development of cancers in a broad range of tissues.Specifications
| C17orf28 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IV36 | |
| HID1 | |
| Full length human recombit protein of human C17orf28 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| HID1 | |
| 17orf28, C17orf28, DMC1, HID-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 283987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction