missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf28 Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501359S
This item is not returnable.
View return policy
Description
C17orf28 Monoclonal antibody specifically detects C17orf28 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| C17orf28 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P24557, P49430 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 24886, 6916 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| TBXAS1 | |
| C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog | |
| Full length human recombinant protein of human TBXAS1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction