Learn More
C17orf37 Monoclonal Antibody (OTI1E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504609
Encompass_Preferred
 View more versions of this product
Product Code. 17532032
  • € 540.87 / 100µL
Description
During the past decade, the role of the ERBB2(neu/HER2) oncogene as an important predictor of patient outcome and response to various therapies in breast cancer has been clearly established. The C35 (C17orf37) is a novel tumor biomarker abundantly expressed in breast cancer. Identification of shared tumor-specific targets is useful in developing broadly applicable therapies. The C35 gene is located on chromosome 17q12, 505nucleotides from the 3′ end of the ERBB2oncogene, the antigenic target for trastuzumab(Herceptin) therapy. The chromosomal arrangement of the genes encoding C35 andERBB2 is tail to tail. The ORF 37 open reading frame encodes a 12-kDa protein of unknown function.
Specifications
Specifications
C17orf37
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BRT3
MIEN1
Full length human recombit protein of human C17orf37 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI1E4
Unconjugated
MIEN1
C17orf37, C35, ORB3, RDX12, XTP4
Mouse
Affinity Chromatography
RUO
84299
-20° C, Avoid Freeze/Thaw Cycles