Learn More
C17orf37 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA504614S
Encompass_Preferred
 View more versions of this product
Product Code. 17502052
  • € 242.00 / 30µL
Description
C17orf37 Monoclonal antibody specifically detects C17orf37 in Human samples. It is validated for Western Blot
Specifications
Specifications
C17orf37
Monoclonal
0.67 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BRT3
Mouse
Affinity chromatography
RUO
84299
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G2
Unconjugated
MIEN1
1810046J19Rik; AI463380; C16H17orf37; C17orf37; C35; HBV XAg-transactivated protein 4; HBV X-transactivated gene 4 protein; MGC14832; MIEN1; Migration and invasion enhancer 1; ORB3; protein C17orf37; protein C17orf37 homolog; Protein C35; RDX12; RGD1306682; XTP4
Full length human recombinant protein of human C17orf37 produced in E.coli.