missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf37 Monoclonal Antibody (OTI3D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800017S
This item is not returnable.
View return policy
Description
C17orf37 Monoclonal antibody specifically detects C17orf37 in Human samples. It is validated for Western BlotSpecifications
| C17orf37 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BRT3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84299 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3D9 | |
| Unconjugated | |
| MIEN1 | |
| 1810046J19Rik; AI463380; C16H17orf37; C17orf37; C35; HBV XAg-transactivated protein 4; HBV X-transactivated gene 4 protein; MGC14832; MIEN1; Migration and invasion enhancer 1; ORB3; protein C17orf37; protein C17orf37 homolog; Protein C35; RDX12; RGD1306682; XTP4 | |
| Full length human recombinant protein of human C17orf37 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction