Learn More
C17orf37 Mouse anti-Human, Clone: OTI1E4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504609
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
During the past decade, the role of the ERBB2(neu/HER2) oncogene as an important predictor of patient outcome and response to various therapies in breast cancer has been clearly established. The C35 (C17orf37) is a novel tumor biomarker abundantly expressed in breast cancer. Identification of shared tumor-specific targets is useful in developing broadly applicable therapies. The C35 gene is located on chromosome 17q12, 505nucleotides from the 3′ end of the ERBB2oncogene, the antigenic target for trastuzumab(Herceptin) therapy. The chromosomal arrangement of the genes encoding C35 andERBB2 is tail to tail. The ORF 37 open reading frame encodes a 12-kDa protein of unknown function.Specifications
| C17orf37 | |
| Monoclonal | |
| Unconjugated | |
| MIEN1 | |
| C17orf37, C35, ORB3, RDX12, XTP4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84299 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1E4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9BRT3 | |
| MIEN1 | |
| Full length human recombit protein of human C17orf37 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.