missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf58 Antibody (OTI6E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46593
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
C17orf58 Monoclonal specifically detects C17orf58 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| C17orf58 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| chromosome 17 open reading frame 58, hypothetical protein LOC284018, MGC138278 | |
| Mouse | |
| 11 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI6E9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| C17orf58 | |
| Full length human recombinant protein of human C17orf58(NP_858041) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 284018 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering