missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C17orf59 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15949054
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15949054 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949054 Supplier Invitrogen™ Supplier No. PA566346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PETDLLAVAEH QALVFGGGPG RTSSEPPAGL RVSGEEETEN VGGANRHPRT SPKTSSCGVV HRPEREALEN EPGPQGTLS Highest antigen sequence identity to the following orthologs - mouse 50%, rat 54%.

As part of the BORC complex may play a role in lysosomes movement and localization at the cell periphery. Associated with the cytosolic face of lysosomes, the BORC complex may recruit ARL8B and couple lysosomes to microtubule plus-end-directed kinesin motor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf59
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BORCS6
Gene Accession No. Q96GS4
Gene Alias 2310047M10Rik; BLOC-1 related complex subunit 6; BLOC-1-related complex subunit 6; BORCS6; C17orf59; LOC497934; Lysosome-dispersing protein; Lyspersin; PRO2472; RIKEN cDNA 2310047M10 gene; similar to hypothetical protein FLJ20014; uncharacterized protein C17orf59; uncharacterized protein C17orf59 homolog
Gene Symbols BORCS6
Host Species Rabbit
Immunogen Recombinant Human C17orf59. Recombinant protein control fragment (Product #RP-105158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54785
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.