missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf62 Monoclonal Antibody (OTI2B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506995S
This item is not returnable.
View return policy
Description
C17orf62 Monoclonal antibody specifically detects C17orf62 in Human samples. It is validated for Western BlotSpecifications
| C17orf62 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15127 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10066 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2B8 | |
| Unconjugated | |
| Scamp2 | |
| C17orf62; chromosome 17 open reading frame 62; CYBC1; Cytochrome b-245 chaperone 1; EROS; Essential for reactive oxygen species protein; HAPSTR1; uncharacterized protein C17orf62 | |
| Full length human recombinant protein of human SCAMP2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction