missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C17orf80 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18086401
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18086401 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18086401 Supplier Invitrogen™ Supplier No. PA5144697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PRETTYQFHS VSQSSSQSLA SLATTFLQEK KAEAQNHNCV PDVKALMESP EGQLSLEPKS DSQFQASHTG CQSPLCSAQR HTPQSPFTNH AAAAGRKTLR SCMGLEWFPE LYPGYLGLGV LPGKPQCWNA MTQKPQ Antigen sequence identity: Mouse 35%; Rat 38%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf80
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C17orf80
Gene Accession No. Q9BSJ5
Gene Alias C17orf80; Cell migration-inducing gene 3 protein; chromosome 17 open reading frame 80; HLC8; HLC-8; Human lung cancer oncogene 8 protein; lung cancer-related protein 8; MIG3; SPEP1; sperm-expressed protein 1; uncharacterized protein C17orf80
Gene Symbols C17orf80
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 358-483. Recombinant protein control fragment (Product #RP-109974).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55028
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.