Learn More
Invitrogen™ C18orf19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562434
Description
Immunogen sequence: QWNVPRTVSR LARRTCLEPH NAGLFGHCQN VKGPLLLYNA ESKVVLVQGP QKQWLHLSAA QCVAKERRPL DAHPPQPGVL RHKQGKQHVS FRRVFSSSAT AQGTP Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 53%.
Specifications
| C18orf19 | |
| Polyclonal | |
| Unconjugated | |
| FAM210A | |
| C18orf19; FAM210A; family with sequence similarity 210 member A; family with sequence similarity 210, member A; HsT2329; Protein FAM210A; uncharacterized protein C18orf19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 125228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96ND0 | |
| FAM210A | |
| Recombinant protein corresponding to Human C18orf19. Recombinant protein control fragment (Product #RP-90489). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.