missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C18orf54 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808319S
This item is not returnable.
View return policy
Description
C18orf54 Monoclonal antibody specifically detects C18orf54 in Human samples. It is validated for Western BlotSpecifications
| C18orf54 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86YS3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84440 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2B2 | |
| Unconjugated | |
| RAB11FIP4 | |
| C18orf54; chromosome 18 open reading frame 54; LAS2; Lung adenoma susceptibility protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction