missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf10, Mouse anti-Human, Clone: 2D4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00056005-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAPSGGWNGVGASLWAALLLGAVALRPAEAVSEPTTVAFDVRPGGVVHSFSHNVGPGDKYTCMFTYASQGGTNEQWQMSLGTSEDHQHFTCTIWRPQGKSYLYFTQFKAEVRGAEIEYAMAYSKAAFERESDVPLKTEEFEVTKTAVAHRPGAFKAELSKLVIVAKASRTELSpecifications
| C19orf10 | |
| Monoclonal | |
| Unconjugated | |
| chromosome 19 open reading frame 10 | |
| EUROIMAGE1875335/IL25/IL27/IL27w/R33729_1/SF20 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence | |
| 2D4 | |
| In 1x PBS, pH 7.4 | |
| BC010129 | |
| C19orf10 | |
| C19orf10 (AAH10129, 1 a.a. ∼ 173 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 56005 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction