Learn More
PeproTech C19orf10 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
€ 219.00 - € 3435.00
Specifications
| Antigen | C19orf10 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17864813
![]() |
PeproTech
500-P259BT-25UG |
25 μg |
€ 219.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17875323
|
PeproTech
500-P259BT-50UG |
50 μg |
€ 275.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17845513
|
Invitrogen
500-P259BT-1MG |
2 x 500 μg | |||||||
|
30054433
|
PeproTech
500-P259BT-500UG |
500 μg |
€ 1836.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine SF-20. Anti-Murine SF-20-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mSF-20 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine SF-20 (500-P259) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mSF-20. Western Blot: To detect mSF-20 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mSF-20 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P259BT-1MG will be provided as 2 x 500 μg
The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. Cytokines are small, soluble proteins with pleiotropic effects on a variety of cell types. Cytokines have a regulatory function over the immune system and mediate aspects of inflammatory response. IL-25 (also called SF20/IL-25) is a secreted bone marrow stroma-derived growth factor cytokine that is derived from Th2 T-cells. IL-25 belongs to the IL-17 cytokine family and is capable of amplifying allergic type inflammatory responses by its actions on other cell types. IL-25 binds to mouse thymic shared antigen-1 and supports lymphoid cell proliferation. Infusion of mice with IL-25 induces IL-4, IL-5, and IL-13 gene expression. IL-25 induced mice also develop epithelial cell hyperplasia, increased mucus secretion, and airway hyperreactivity, suggesting that IL-25 may be an important mediator of allergic disease via production of IL-4, IL-5, IL-13 and eotaxin.Specifications
| C19orf10 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Mouse | |
| Q9CPT4 | |
| 28106 | |
| E.coli-derived, 15.8 kDa Recombinant Murine SF-20. | |
| Antigen affinity chromatography | |
| MYDGF |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| C19orf10; CS010; D17Wsu104e; EUROIMAGE1875335; IL 25; IL 27; Il25; IL27; IL27w; interleukin 25; interleukin 27 working designation; Interleukin25; interleukin-25; LOC501282; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; R33729 1; R33729_1; R337291; SF20; similar to lymphocyte antigen 6 complex, locus E ligand; stromal cell-derived growth factor SF20; UPF0556 protein C19orf10; UPF0556 protein C19orf10 homolog | |
| MYDGF | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.