missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf44 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98139AF350
This item is not returnable.
View return policy
Description
C19orf44 Polyclonal antibody specifically detects C19orf44 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| C19orf44 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84167 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| chromosome 19 open reading frame 44, FLJ21742, FLJ22492, hypothetical protein LOC84167 | |
| Produced in rabbits immunized with E. coli-derived Human C19orf44 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction